Merck
  • Active, phosphorylated fingolimod inhibits histone deacetylases and facilitates fear extinction memory.
Active, phosphorylated fingolimod inhibits histone deacetylases and facilitates fear extinction memory.
Nature neuroscience (2014-05-27)
Nitai C Hait, Laura E Wise, Jeremy C Allegood, Megan O'Brien, Dorit Avni, Thomas M Reeves, Pamela E Knapp, Junyan Lu, Cheng Luo, Michael F Miles, Sheldon Milstien, Aron H Lichtman, Sarah Spiegel
ABSTRACT
FTY720 (fingolimod), an FDA-approved drug for treatment of multiple sclerosis, has beneficial effects in the CNS that are not yet well understood, independent of its effects on immune cell trafficking. We show that FTY720 enters the nucleus, where it is phosphorylated by sphingosine kinase 2 (SphK2), and that nuclear FTY720-P binds and inhibits class I histone deacetylases (HDACs), enhancing specific histone acetylations. FTY720 is also phosphorylated in mice and accumulates in the brain, including the hippocampus, inhibits HDACs and enhances histone acetylation and gene expression programs associated with memory and learning, and rescues memory deficits independently of its immunosuppressive actions. Sphk2(-/-) mice have lower levels of hippocampal sphingosine-1-phosphate, an endogenous HDAC inhibitor, and reduced histone acetylation, and display deficits in spatial memory and impaired contextual fear extinction. Thus, sphingosine-1-phosphate and SphK2 play specific roles in memory functions and FTY720 may be a useful adjuvant therapy to facilitate extinction of aversive memories.
Sigma-Aldrich
Sphingosine 1-phosphate, ≥95%, powder
Sigma-Aldrich
Sphingosine 1-phosphate, ≥98.0% (TLC)
Sigma-Aldrich
FTY720, ≥98% (HPLC)
Sigma-Aldrich
Anti-Histone H4 Antibody, Upstate®, from rabbit
Sigma-Aldrich
D-Sphingosine, synthetic
Sigma-Aldrich
Anti-acetyl-Histone H4 (Lys16) Antibody, Upstate®, from rabbit
Sigma-Aldrich
Anti-Histone H2B Antibody, Upstate®, from rabbit
Sigma-Aldrich
Anti-acetyl-Histone H3 (Lys23) Antibody, serum, Upstate®
Sigma-Aldrich
Anti-acetyl-Histone H3 (Lys18) Antibody, serum, Upstate®