Merck
Brønsted-acid and Brønsted-base catalyzed Diels-Alder reactions.
Organic & biomolecular chemistry (2008-09-20)
Juan Shen, Choon-Hong Tan
ABSTRACT
The enantioselective Diels-Alder reaction is one of the most important reactions for the synthesis of complex molecules. It provides access to chiral six-membered carbocyclic compounds containing up to four stereogenic centers in a single step. Asymmetric catalysis in the Diels-Alder reaction has mainly been realized using chiral Lewis acids. In this perspective, we describe several cases of chiral Brønsted-acid and Brønsted-base catalyzed Diels-Alder reactions, providing an overview of this rapidly growing field.
Sigma-Aldrich
Ethyl 3-benzoylacrylate, technical grade, 92%