Merck
  • Structural elucidation and synthesis of 3-hydroxybenzene-1,2-dicarbaldehyde from astigmatid mites.
Structural elucidation and synthesis of 3-hydroxybenzene-1,2-dicarbaldehyde from astigmatid mites.
Bioscience, biotechnology, and biochemistry (2002-01-05)
T Sakata, Y Kuwahara
ABSTRACT
The structure of a novel aromatic compound contained in the unidentified Rhizoglyhus mite (Acaridae: Astigmata) was elucidated, without its isolation, to be 3-hydroxybenzene-1,2-dicarbaldehyde (tentatively named gamma-acaridial) by a combination of GC/MS and GC/FT-IR together with knowledge of related mite compounds. The structure was confirmed by a 5-step synthesis, starting from methyl 3-hydroxybenzoate, in a 15% overall yield. The compound is widely distributed not only among Astigmata but also among Oribatida, although its biological function is obscure at present.
Sigma-Aldrich
Methyl 3-hydroxybenzoate, 99%