Merck
  • [Comparison of the modifying action of actinomycin D and xanthobine on mouse survival after 2-stage irradiation].
[Comparison of the modifying action of actinomycin D and xanthobine on mouse survival after 2-stage irradiation].
Radiobiologiia (1984-07-01)
V A Volchkov, N Ia Mikhaĭlova
ABSTRACT
It was shown that xanthobin and actinomycin D decreased significantly, to an approximately the same degree, the survival of mice at 6- and 9-hour intervals between two whole-body exposures to X-rays (6 + 6 Gy).