Merck
  • Changes in hemostatic parameters after oral and transdermal hormone therapy in postmenopausal women.
Changes in hemostatic parameters after oral and transdermal hormone therapy in postmenopausal women.
Gynecological endocrinology : the official journal of the International Society of Gynecological Endocrinology (2010-10-29)
Ireneusz Polac, Marta Borowiecka, Agnieszka Wilamowska, Michal Bijak, Michal B Ponczek, Pawel Nowak
ABSTRACT
Hormone therapy (HT) can be prothrombotic risk factor. We compared the effects of oral HT (o-HT) and transdermal HT (t-HT) on the kinetic of clot formation and fibrinolysis in postmenopausal women after 6 months HT using a multiparameter test. We observed that after HT, the level of fibrinogen was higher than in controls (Fg 3.12 g/l vs. 4.24 g/l (o-HT); 3,7 g/l (t-HT); p < 0.001) and values of velocity of polymerization in o-HT group were increased (95.84 mOD/min vs. 146.50 mOD/min, p < 0.001) compared to controls. Maximum absorbance of formed clots was higher in o-HT group (0.279 vs. 0.312, p < 0.001) than in controls, but in t-HT group was lowest (0.268). Fibrin lysis half-time increased in both HT groups (controls 17.16 min vs. 31.43 min (o-HT); 23.34 min (t-HT) p < 0.001) compared to values in controls. The results of our study show that o-HT caused the changes in clot formation and fibrinolysis than t-HT in postmenopausal women. The increased level of fibrinogen and its accelerated kinetics of polymerization as well as a lower rate of clot lysis may partly explain the increase in venous thrombosis and cardiovascular events reported after the use of HT, especially the oral form of that.
Product Description
Dydrogesterone, European Pharmacopoeia (EP) Reference Standard