Merck
  • Determination of leakage from antibody adsorbent: composition analysis and pH effect.
Determination of leakage from antibody adsorbent: composition analysis and pH effect.
Biomedical chromatography : BMC (2013-05-18)
Lan Zhao, Yongdong Liu, Yinjue Wang, Yongdong Huang, Xiunan Li, Yan Li, Guanghui Ma, Zhiguo Su
ABSTRACT
To study the leakage at different solution pH values, IgG Sepharose 6FF®, a commercially available immunoadsorbent, was used as a model. The leaked substance consists of three parts: (1) ligands and its fragments; (2) ligands plus matrix fragments in which ligands are chemically attached to the adsorbent matrix; and (3) matrix fragments. Buffer solution pH values had a great effect on both the kinetics and the amount of ligand leakage. Cross-linking of the adsorbent matrix could reduce both matrix leakage and antibody leakage at pH 3.0, but its effect was limited at pH 11.0 for ligand leakage.
Sigma-Aldrich
Agarose, High EEO, for molecular biology
Sigma-Aldrich
Agarose, Type II-A, Medium EEO
Sigma-Aldrich
Agarose, Type IV, Special High EEO
Sigma-Aldrich
Agarose, BioReagent, for molecular biology, Wide range/Standard 3:1
Sigma-Aldrich
Agarose, Type I-A, low EEO
Sigma-Aldrich
Agarose, for molecular biology
Sigma-Aldrich
Sepharose 6B, 6% Beaded Agarose, 45-165 μm (wet), fractionation range 10,000-1,000,000 Da (dextrans)
Sigma-Aldrich
Agarose, High EEO
Sigma-Aldrich
Agarose, High Gelling Temperature
Sigma-Aldrich
Agarose, For pulsed field electrophoresis running gel
Sigma-Aldrich
Agarose, BioReagent, for molecular biology, low EEO
Sigma-Aldrich
Agarose, Medium EEO, for molecular biology
Sigma-Aldrich
Agarose, Low EEO, for Immunoelectrophoresis
Sigma-Aldrich
Agarose, Low EEO
Sigma-Aldrich
Agarose, Special High EEO
Sigma-Aldrich
Agarose, Ultra-low Gelling Temperature, molecular biology grade
Sigma-Aldrich
Agarose, Wide range, for molecular biology
Sigma-Aldrich
Agarose, Type I, low EEO
Sigma-Aldrich
Agarose, Ultra-low Gelling Temperature
Sigma-Aldrich
Agarose, Vetec, reagent grade
Sigma-Aldrich
Sepharose 4B, 45-165 μm bead diameter