Merck
  • Mass, IR, electronic and EPR spectral studies on transition metal complexes with a new tetradentate 12-membered new macrocyclic ligand.
Mass, IR, electronic and EPR spectral studies on transition metal complexes with a new tetradentate 12-membered new macrocyclic ligand.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2004-10-13)
Sulekh Chandra, Lokesh Kumar Gupta
ABSTRACT
Complexes of Cr(III), Co(II), Ni(II) and Cu(II) containing a novel macrocyclic tetradentate nitrogen donor (N4) ligand prepared via reaction of 2,3-hexanedione and ethylenediamine has been prepared and characterized. The newly synthesized ligand (L) and its complexes have been characterized on the basis of elemental analysis, molar conductance, magnetic moment susceptibility, EI-Mass, IR, Electronic and EPR spectral studies. The complexes are of high-spin type and four coordinated tetrahedral, five coordinated square pyramidal and six coordinated octahedral/tetragonal geometries. The ligand (L) and its soluble transition metal complexes have also been screened against different bacteria and plant pathogenic fungi in vitro.
Sigma-Aldrich
2,3-Hexanedione, technical grade, 90%