Merck
  • Pharmacokinetics of methylphenidate in the rat using single-ion monitoring GLC-mass spectrometry.
Pharmacokinetics of methylphenidate in the rat using single-ion monitoring GLC-mass spectrometry.
Journal of pharmaceutical sciences (1977-06-01)
J Gal, B J Hodshon, C Pintauro, B L Flamm, A K Cho
PMID874790
ABSTRACT
A GLC-mass spectrometric assay for methylphenidate in biological fluids was developed using the ethyl ester homolog of the drug as the internal standard. The procedure has a lower level of sensitivity of 1.2 ng/ml and is based on GLC-mass spectrometic monitoring of the m/e 180 ion common to the mass spectra of the N-trifluoroacetyl derivatives of the drug and internal standard. The brain and plasma levels of methylphenidate in rats were determined after intravenous administration of 0.5 mg/kg of the drug. The two-compartment open pharmacokinetic model fit the data.
Product Description
Supelco
Trifluoroacetic anhydride, for GC derivatization, LiChropur, ≥99.0% (GC)