- Protective effects of 5-iodo-6-amino-1,2-benzopyrone, an inhibitor of poly(ADP-ribose) synthetase against peroxynitrite-induced glial damage and stroke development.
Protective effects of 5-iodo-6-amino-1,2-benzopyrone, an inhibitor of poly(ADP-ribose) synthetase against peroxynitrite-induced glial damage and stroke development.
European journal of pharmacology (1998-08-28)
M Endres, G S Scott, A L Salzman, E Kun, M A Moskowitz, C Szabó
PMID9721031
ABSTRACT
Peroxynitrite triggers DNA single-strand breakage, which activates the nuclear enzyme poly(ADP-ribose) synthetase (PARS). Activation of PARS depletes its substrate, NAD+, slowing the rate of glycolysis, electron transport, and ATP formation, resulting in cell necrosis. Here, we demonstrate that inhibition of PARS with the novel, potent PARS inhibitor 5-iodo-6-amino-1,2-benzopyrone (INH2BP) protects against peroxynitrite-induced cell death (as measured by measurement of mitochondrial respiration and release of lactate dehydrogenase) in C6 glioma cells in vitro, and in a murine stroke model in vivo. Inhibition of PARS with INH2BP may represent a novel approach for the experimental therapy of stroke.