Merck
Multiple anthelmintic resistance in Trichostrongylus colubriformis.
Australian veterinary journal (1986-02-01)
K M Dash
ABSTRACT
Following the failure of anthelmintic treatment to control an outbreak of trichostrongylosis in sheep, multiple resistance to levamisole and oxfendazole was confirmed in field strains of Trichostrongylus colubriformis at the CSIRO Pastoral Research Laboratory, Armidale. Resistance in Trichostrongylus spp to levamisole was also confirmed on an adjoining farm. From the results of an experiment where lambs were treated at the recommended dose rate with one or both anthelmintics, it was estimated that 32% of T. colubriformis were resistant to levamisole, 19% to oxfendazole, and 12% to both drugs. Simultaneous administration of levamisole and oxfendazole resulted in an additive anthelmintic effect. Naphthalophos (36.6 to 51.2 mg/kg) was 93% efficient against the multiple resistant strain. The similar histories of anthelmintic usage at the CSIRO Pastoral Research Laboratory and on the northern tablelands of New South Wales generally, suggest that multiple anthelmintic resistance in T. colubriformis may soon emerge as a problem on the northern tablelands.