- Protective effect of vitamins against trichothecene toxicity towards Saccharomyces cerevisiae.
Protective effect of vitamins against trichothecene toxicity towards Saccharomyces cerevisiae.
Experientia (1987-08-15)
B Yagen, S Halevy
PMID3305064
ABSTRACT
Several trichothecene mycotoxins were shown to inhibit the growth of Saccharomyces cerevisiae. This effect was most pronounced with the macrocyclic trichothecenes, especially verrucarin A. Much less growth inhibition was observed with T-2 toxin. Verrucarol, diacetoxyscirpenol, acetyl T-2 toxin, HT-2 toxin, T-2 tetraol and neosolaniol were inactive at a concentration of 75 micrograms of toxin per disc. Incubation of S. cerevisiae with verrucarin A together with vitamins resulted in a decrease in toxicity. Pyridoxine-HCl, Ca-pantothenate, thiamine-HCl and alpha-tocopheryl acetate were amongst the most potent of the vitamins tested which reversed growth inhibition, overcoming the inhibitory potential of the toxins.