- Inhibition of vasopressin-stimulated flank marking behavior by V1-receptor antagonists.
Inhibition of vasopressin-stimulated flank marking behavior by V1-receptor antagonists.
Flank marking, a form of olfactory communication displayed by hamsters, is dependent upon vasopressin-sensitive neurons in the anterior hypothalamus. In the present study two vasopressin type-1 (V1) receptor antagonists, d(CH2)5Tyr(Me)AVP and dPTyr(Me)AVP were tested for their ability to block flank marking stimulated by the microinjection of arginine vasopressin (AVP) into the anterior hypothalamus. Dose-response curves were established for AVP and flank marking in the presence or absence of different concentrations of each antagonist. DPTyr(Me)AVP was microinjected into the anterior hypothalamus 1 h before the microinjection of AVP while d(CH2)5Tyr(Me)AVP and AVP were prepared together and delivered as a single microinjection. This procedure was necessary because dPTyr(Me)AVP, but not d(CH2)5Tyr(Me)AVP, had agonist activity when initially injected into the anterior hypothalamus in concentrations ranging from 0.90-900 microM. The ED50 values (microM) for dPTyr(Me)AVP and AVP were 17.9 and 0.90, respectively. The initial agonist activity of dPTyr(Me)AVP was always followed by blocker activity. Both V1-receptor antagonists caused a dose-dependent decrease in AVP-stimulated flank marking. Maximal inhibition of AVP-stimulated flank marking was produced with approximately 1.0 mM of either antagonist. Both antagonists blocked AVP-stimulated flank marking behavior for over 12 h following their microinjection.