Merck
  • Targeting BCL-2 with the BH3 mimetic ABT-199 in estrogen receptor-positive breast cancer.
Targeting BCL-2 with the BH3 mimetic ABT-199 in estrogen receptor-positive breast cancer.
Cancer cell (2013-07-13)
François Vaillant, Delphine Merino, Lily Lee, Kelsey Breslin, Bhupinder Pal, Matthew E Ritchie, Gordon K Smyth, Michael Christie, Louisa J Phillipson, Christopher J Burns, G Bruce Mann, Jane E Visvader, Geoffrey J Lindeman
ABSTRACT
The prosurvival protein BCL-2 is frequently overexpressed in estrogen receptor (ER)-positive breast cancer. We have generated ER-positive primary breast tumor xenografts that recapitulate the primary tumors and demonstrate that the BH3 mimetic ABT-737 markedly improves tumor response to the antiestrogen tamoxifen. Despite abundant BCL-XL expression, similar efficacy was observed with the BCL-2 selective inhibitor ABT-199, revealing that BCL-2 is a crucial target. Unexpectedly, BH3 mimetics were found to counteract the side effect of tamoxifen-induced endometrial hyperplasia. Moreover, BH3 mimetics synergized with phosphatidylinositol 3-kinase (PI3K)/mammalian target of rapamycin (mTOR) inhibitors in eliciting apoptosis. Importantly, these two classes of inhibitor further enhanced tumor response in combination therapy with tamoxifen. Collectively, our findings provide a rationale for the clinical evaluation of BH3 mimetics in therapy for breast cancer.
Sigma-Aldrich
Hexokinase from Saccharomyces cerevisiae, Type III, lyophilized powder, ≥25 units/mg protein (biuret)
Sigma-Aldrich
Hexokinase from Saccharomyces cerevisiae, Type F-300, lyophilized powder, ≥130 units/mg protein (biuret)
Sigma-Aldrich
Hexokinase from Saccharomyces cerevisiae, lyophilized powder, ≥350 units/mg protein, Protein ≥10 % by biuret