Merck
  • Quantification of globotriaosylsphingosine in plasma and urine of fabry patients by stable isotope ultraperformance liquid chromatography-tandem mass spectrometry.
Quantification of globotriaosylsphingosine in plasma and urine of fabry patients by stable isotope ultraperformance liquid chromatography-tandem mass spectrometry.
Clinical chemistry (2012-12-15)
Henrik Gold, Mina Mirzaian, Nick Dekker, Maria Joao Ferraz, Johan Lugtenburg, Jeroen D C Codée, Gijs A van der Marel, Herman S Overkleeft, Gabor E Linthorst, Johanna E M Groener, Johannes M Aerts, Ben J H M Poorthuis
ABSTRACT
Biochemical markers that accurately reflect the severity and progression of disease in patients with Fabry disease and their response to treatment are urgently needed. Globotriaosylsphingosine, also called lysoglobotriaosylceramide (lysoGb3), is a promising candidate biomarker. We synthesized lysoGb3 and isotope-labeled [5,6,7,8,9] (13)C5-lysoGb3 (internal standard). After addition of the internal standard to 25 μL plasma or 400 μL urine from patients with Fabry disease and healthy controls, samples were extracted with organic solvents and the lysoGb3 concentration was quantified by UPLC-ESI-MS/MS (ultraperformance liquid chromatography-electrospray ionization-tandem mass spectrometry). Calibration curves were constructed with control plasma and urine supplemented with lysoGb3. In addition to lysoGb3, lyso-ene-Gb3 was quantified. Quantification was achieved by multiple reaction monitoring of the transitions m/z 786.4 > 282.3 [M+H](+) for lysoGb3, m/z 791.4 > 287.3 [M+H](+) for [5,6,7,8,9] (13)C5-lysoGb3, and 784.4 > 280.3 [M+H](+) for lyso-ene-Gb3. The mean (SD) plasma lysoGb3 concentration from 10 classically affected Fabry hemizygotes was 94.4 (25.8) pmol/mL (range 52.7-136.8 pmol/mL), from 10 classically affected Fabry heterozygotes 9.6 (5.8) pmol/mL (range 4.1-23.5 pmol/mL), and from 20 healthy controls 0.4 (0.1) pmol/mL (range 0.3-0.5 pmol/mL). Lyso-ene-Gb3 concentrations were 10%-25% of total lysoGb3. The urine concentration of lysoGb3 was 40-480 times lower than in corresponding plasma samples. Lyso-ene-Gb3 concentrations in urine were comparable or even higher than the corresponding lysoGb3 concentrations. This assay for the quantification of lysoGb3 and lyso-ene-Gb3 in human plasma and urine samples will be an important tool in the diagnosis of Fabry disease and for monitoring the effect of enzyme replacement therapy in patients with Fabry disease.
Sigma-Aldrich
Globotriaosylsphingosine from porcine blood