Merck
  • Simulation of the hepatic metabolism of stilbene and its tricyclic derivatives by Fenton and Ruff reagents: models for cytochrome P-450 activation of chemical carcinogens.
Simulation of the hepatic metabolism of stilbene and its tricyclic derivatives by Fenton and Ruff reagents: models for cytochrome P-450 activation of chemical carcinogens.
Biopharmaceutics & drug disposition (1990-01-01)
S Zbaida, R Kariv
ABSTRACT
Reactions of trans-stilbene, cis-stilbene, 5H-dibenzo [a,d] cyclo-heptene 5-one and 5H-dibenz [b,f] azepine (iminostilbene) with Fenton reagent [Fe (II)/H2O2] clearly simulate their hepatic metabolism. Expoxidation on the corresponding ethylenic linkage was found to be a common pathway of these compounds. Epoxides of trans-stilbene, cis-stilbene, and 5H-dibenzo[a,d]cycloheptene 5-one were further oxidized to dihydrodiols, alpha-hydroxyketones, diketones, and finally cleavage of the ethylenic bonds to the formation of the corresponding aldehydes. However, the unstable epoxide of iminostilbene gave 9-acridinecarbaldehyde that is further oxidized to 9-acridone. Reaction of both trans- and cis-stilbene with Ruff reagent [Fe III)/H2O2] gave the same oxidative products to that obtained from Fenton reagent. The radical scavenger 2,6 bis (1,1-dimethylethyl)-4-methyl phenol (BHT) decreases the total yield conversion and increases the formation ratio of both cis-epoxide and d,l-hydrobenzoin from cis-stilbene.
Sigma-Aldrich
5H-Dibenz[b,f]azepine, 97%
Pricing and availability is not currently available.
Supelco
Iminostilbene, Pharmaceutical Secondary Standard; Certified Reference Material
Pricing and availability is not currently available.
Dibenzosuberone, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.