Merck
  • Total syntheses of mitragynine, paynantheine and speciogynine via an enantioselective thiourea-catalysed Pictet-Spengler reaction.
Total syntheses of mitragynine, paynantheine and speciogynine via an enantioselective thiourea-catalysed Pictet-Spengler reaction.
Chemical communications (Cambridge, England) (2012-11-15)
Isabel P Kerschgens, Elise Claveau, Martin J Wanner, Steen Ingemann, Jan H van Maarseveen, Henk Hiemstra
ABSTRACT
The pharmacologically interesting indole alkaloids (-)-mitragynine, (+)-paynantheine and (+)-speciogynine were synthesised in nine steps from 4-methoxytryptamine by a route featuring (i) an enantioselective thiourea-catalysed Pictet-Spengler reaction, providing the tetrahydro-β-carboline ring and (ii) a Pd-catalysed Tsuji-Trost allylic alkylation, closing the D-ring.
Product Description
Supelco
Mitragynine solution, 100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®