Merck
  • Microbial degradation and toxicity of hexahydro-1,3,5-trinitro-1,3,5-triazine.
Microbial degradation and toxicity of hexahydro-1,3,5-trinitro-1,3,5-triazine.
Journal of microbiology and biotechnology (2012-10-19)
Muhammad Imran Khan, Jaejin Lee, Joonhong Park
ABSTRACT
In the present work, current knowledge on the potential fate, microbial degradation, and toxicity of hexahydro- 1,3,5-trinitro-1,3,5-triazine (RDX) was thoroughly reviewed, focusing on the toxicological assessment of a variety of potential RDX degradation pathways in bacteria and fungi. The present review on microbial degradation pathways and toxicities of degradation intermediates suggests that, among aerobic RDX degradation pathways, the one via denitration may be preferred in a toxicological perspective, and that among anaerobic pathways, those forming 4- nitro-2,4-diazabutanal (NDAB) via ring cleavage of 1-nitroso- 3,5-dinitro-1,3,5-triazinane (MNX) may be toxicologically advantageous owing to its potential mineralization under partial or complete anoxic conditions. These findings provide important information on RDX-degrading microbial pathways, toxicologically most suitable to be stimulated in contaminated fields.
Product Description
Supelco
RDX solution, 1000 μg/mL in acetonitrile, ampule of 1.2 mL, certified reference material, Cerilliant®