Merck
  • Methyl jasmonate and yeast extract stimulate mitragynine production in Mitragyna speciosa (Roxb.) Korth. shoot culture.
Methyl jasmonate and yeast extract stimulate mitragynine production in Mitragyna speciosa (Roxb.) Korth. shoot culture.
Biotechnology letters (2012-06-21)
Juraithip Wungsintaweekul, Jutarat Choo-Malee, Tossaton Charoonratana, Niwat Keawpradub
ABSTRACT
Mitragynine is a pharmacologically-active terpenoid indole alkaloid found in Mitragyna speciosa leaves. Treatment with methyl jasmonate (10 μM) for 24 h and yeast extract (0.1 mg/ml) for 12 h were the optimum conditions of elicitation of mitragynine accumulation in a M. speciosa shoot culture. The former elicitor gave 0.11 mg mitragynine/g dry wt. Tryptophan decarboxylase and strictosidine synthase mRNA levels were enhanced in accordance with mitragynine accumulation.
Product Description
Supelco
Mitragynine solution, 100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Pricing and availability is not currently available.