Merck
  • Triterpene hexahydroxydiphenoyl esters and a quinic acid purpurogallin carbonyl ester from the leaves of Castanopsis fissa.
Triterpene hexahydroxydiphenoyl esters and a quinic acid purpurogallin carbonyl ester from the leaves of Castanopsis fissa.
Phytochemistry (2011-08-13)
Yong-Lin Huang, Takaaki Tsujita, Takashi Tanaka, Yosuke Matsuo, Isao Kouno, Dian-Peng Li, Gen-ichiro Nonaka
ABSTRACT
Triterpene hexahydroxydiphenoyl (HHDP) esters have only been isolated from Castanopsis species, and the distribution of these esters in nature is of chemotaxonomical interest. In this study, the chemical constituents of the leaves of Castanopsis fissa were examined in detail to identify and isolate potential HHDP esters. Together with 53 known compounds, 3,4-di-O-galloyl-1-O-purpurogallin carbonyl quinic acid (1) and 3,24-(S)-HHDP-2α,3β,23,24-tetrahydroxytaraxastan-28,20β-olide (2) were isolated and their structures were elucidated by spectroscopic and chemical methods. The polyphenols of the leaves were mainly composed of galloyl quinic acids, triterpenes HHDP esters, ellagitannins and flavonol glycosides. In particular, the isolation yields of 1,3,4-trigalloyl quinic acid and compound 2 were 1.53% and 0.27%, respectively, from the fresh leaves. The presence of lipid soluble HHDP esters of oleanane-type triterpenes as one of the major metabolites is an important chemotaxonomical discovery. Lipase inhibition activities and ORAC values of the major constituents were compared. The triterpene HHDP ester showed moderate lipase inhibition activity and myricitrin gave the largest ORAC value.