Merck
  • Chemical composition and antimicrobial activity of the essential oil of Rosemary.
Chemical composition and antimicrobial activity of the essential oil of Rosemary.
Environmental toxicology and pharmacology (2011-07-27)
Yang Jiang, Nan Wu, Yu-Jie Fu, Wei Wang, Meng Luo, Chun-Jian Zhao, Yuan-Gang Zu, Xiao-Lei Liu
ABSTRACT
The composition of the essential oil of Rosemary was analyzed by gas chromatography-mass spectrometry (GC-MS). 22 components, which constitute 97.41% of the oil, were identified. The major constituents were 1,8-Cineole (26.54%) and α-Pinene (20.14%). Minimum inhibitory concentrations (MICs), minimal bactericidal concentration (MBC) and time-kill dynamic processes against three Gram-positive bacteria (Staphylococcus epidermidis, Staphylococcus aureus and Bacillus subtilis), three Gram-negative bacteria (Proteus vulgaris, Pseudomonas aeruginosa and Escherichia coli) and two fungi (Candida albicans and Aspergillus niger) were determined for the oil, 1,8-Cineole and α-Pinene. The oil showed pronounced antibacterial and antifungal activity than 1,8-Cineole and α-Pinene against all of the tested microbes. Furthermore, the survival rates and morphological changes of S. aureus after treatment with different concentrations of the essential oil were assessed by flow cytometry (FCM) and atomic force microscopy (AFM).
Sigma-Aldrich
Rosemary oil, FG
Sigma-Aldrich
Rosemary oil, Certified organic (NOP)