Merck
  • A simple and rapid method for measurement of (10)B-para-boronophenylalanine in the blood for boron neutron capture therapy using fluorescence spectrophotometry.
A simple and rapid method for measurement of (10)B-para-boronophenylalanine in the blood for boron neutron capture therapy using fluorescence spectrophotometry.
Journal of radiation research (2009-06-10)
Genro Kashino, Satoshi Fukutani, Minoru Suzuki, Yong Liu, Kenji Nagata, Shin-Ichiro Masunaga, Akira Maruhashi, Hiroki Tanaka, Yoshinori Sakurai, Yuko Kinashi, Noriko Fujii, Koji Ono
ABSTRACT
Background and Purpose; (10)B deriving from (10)B-para-boronophenylalanine (BPA) and (10)B-borocaptate sodium (BSH) have been detected in blood samples of patients undergoing boron neutron capture therapy (BNCT) using prompt gamma ray spectrometer or Inductively Coupled Plasma (ICP) method, respectively. However, the concentration of each compound cannot be ascertained because boron atoms in both molecules are the target in these assays. Here, we propose a simple and rapid method to measure only BPA by detecting fluorescence based on the characteristics of phenylalanine. Material and Methods; (10)B concentrations of blood samples from human or mice were estimated by the fluorescence intensities at 275 nm of a BPA excited by light of wavelength 257 nm using a fluorescence spectrophotometer. Results; The relationship between fluorescence to increased BPA concentration showed a positive linear correlation. Moreover, we established an adequate condition for BPA measurement in blood samples containing BPA, and the estimated (10)B concentrations of blood samples derived from BPA treated mice were similar between the values obtained by our method and those by ICP method. Conclusion; This new assay will be useful to estimate BPA concentration in blood samples obtained from patients undergoing BNCT especially in a combination use of BSH and BPA.
Sigma-Aldrich
4-Borono-L-phenylalanine, ≥95.0% (HPLC)