Merck
  • Effect of Chia oil (Salvia Hispanica) rich in omega-3 fatty acids on the eicosanoid release, apoptosis and T-lymphocyte tumor infiltration in a murine mammary gland adenocarcinoma.
Effect of Chia oil (Salvia Hispanica) rich in omega-3 fatty acids on the eicosanoid release, apoptosis and T-lymphocyte tumor infiltration in a murine mammary gland adenocarcinoma.
Prostaglandins, leukotrienes, and essential fatty acids (2007-07-10)
C E Espada, M A Berra, M J Martinez, A R Eynard, M E Pasqualini
ABSTRACT
We investigated the effects of certain dietary polyunsaturated fatty acids (PUFAs) and related eicosanoids on the growth and metastasis formation of a murine mammary gland adenocarcinoma. Salvia hispanica (ChO) and Carthamus tinctorius (SaO) vegetable oil sources of omega-3 and -6 PUFAs and a commercial diet as control (CO), were used. We analysed fatty acids of neoplastic cells (NC) membranes by GLC; the eicosanoids 12- HETE and 12-HHT (LOX and COX metabolites) by HPLC and apoptosis and T-lymphocyte infiltration by flow cytometry and microscopy. NC from ChO groups showed lower levels of arachidonic acid and of both eicosanoids compared to SaO and CO (p<0.05). The ChO diet decreased the tumor weight and metastasis number (p<0.05). Apoptosis and T-lymphocyte infiltration were higher and mitosis decreased with respect to the other diets (p<0.05). Present data showed that ChO, an ancient and almost unknown source of omega-3, inhibits growth and metastasis in this tumor model.
Sigma-Aldrich
12(S)-Hydroxy-(5Z,8E,10E)-heptadecatrienoic acid, ≥93% (HPLC), ethanol solution