Merck
  • Dynamic kinetic resolution and desymmetrization processes: a straightforward methodology for the enantioselective synthesis of piperidines.
Dynamic kinetic resolution and desymmetrization processes: a straightforward methodology for the enantioselective synthesis of piperidines.
Chemistry (Weinheim an der Bergstrasse, Germany) (2006-07-20)
Mercedes Amat, Oriol Bassas, Núria Llor, Margalida Cantó, Maria Pérez, Elies Molins, Joan Bosch
ABSTRACT
A straightforward procedure for the synthesis of enantiopure polysubstituted piperidines is reported. It involves the direct generation of chiral non-racemic oxazolo[3,2-a]piperidone lactams that already incorporate carbon substituents on the heterocyclic ring and the subsequent removal of the chiral auxiliary. The key step is a cyclocondensation reaction of (R)-phenylglycinol or other amino alcohols with racemic or prochiral delta-oxo (di)acid derivatives in highly stereoselective processes involving dynamic kinetic resolution and/or desymmetrization of diastereotopic or enantiotopic ester groups.
Sigma-Aldrich
N-(2-Hydroxyethyl)aniline, 98%