Merck
  • Identification of the cytochrome P450 enzymes responsible for the omega-hydroxylation of phytanic acid.
Identification of the cytochrome P450 enzymes responsible for the omega-hydroxylation of phytanic acid.
FEBS letters (2006-06-20)
J C Komen, R J A Wanders
ABSTRACT
Patients suffering from Refsum disease have a defect in the alpha-oxidation pathway which results in the accumulation of phytanic acid in plasma and tissues. Our previous studies have shown that phytanic acid is also a substrate for the omega-oxidation pathway. With the use of specific inhibitors we now show that members of the cytochrome P450 (CYP450) family 4 class are responsible for phytanic acid omega-hydroxylation. Incubations with microsomes containing human recombinant CYP450s (Supersomes) revealed that multiple CYP450 enzymes of the family 4 class are able to omega-hydroxylate phytanic acid with the following order of efficiency: CYP4F3A>CYP4F3B>CYP4F2>CYP4A11.