Merck
  • Eosinophil trans-basement membrane migration induced by interleukin-8 and neutrophils.
Eosinophil trans-basement membrane migration induced by interleukin-8 and neutrophils.
American journal of respiratory cell and molecular biology (2006-02-04)
Izumi Kikuchi, Shinya Kikuchi, Takehito Kobayashi, Koichi Hagiwara, Yoshio Sakamoto, Minoru Kanazawa, Makoto Nagata
ABSTRACT
Neutrophilic inflammation observed with severe asthma is often associated with interleukin-8 (IL-8). Neutrophils can secrete a variety of mediators that may augment the migration of eosinophils. We have reported a positive correlation between the concentrations of neutrophils and eosinophils in sputum from subjects with severe asthma, suggesting a possible role of neutrophils in regulating eosinophilic inflammation. The aim of this study was to investigate whether neutrophils stimulated with IL-8 modify the trans-basement membrane migration (TBM) of eosinophils. Eosinophils and neutrophils were isolated from peripheral blood drawn from healthy donors or subjects with mild asthma. The TBM of eosinophils in response to IL-8 was evaluated in the presence or absence of neutrophils using the chambers with a Matrigel-coated transwell insert. Neither IL-8 alone nor the presence of neutrophils alone induced the TBM of eosinophils. However, when eosinophils were coincubated with neutrophils and stimulated with IL-8, the TBM of eosinophils was significantly augmented. This augmented TBM of eosinophils was inhibited by a matrix metalloproteinase-9 inhibitor, a leukotriene B4 receptor antagonist, platelet-activating factor antagonists, or an anti-TNF-alpha monoclonal antibodies. These results suggest that neutrophils migrated in response to IL-8 may lead eosinophils to accumulate in the airways of asthma and possibly aggravate this disease.
Sigma-Aldrich
WEB2086, ≥98% (HPLC)