Merck
  • Synthesis and biological properties of new alpha-methylene-gamma-butyrolactones and alpha,beta-unsaturated delta-lactones.
Synthesis and biological properties of new alpha-methylene-gamma-butyrolactones and alpha,beta-unsaturated delta-lactones.
European journal of medicinal chemistry (2005-12-22)
Francesca Cateni, Jelena Zilic, Marina Zacchigna, Paolo Bonivento, Fabiana Frausin, Vito Scarcia
ABSTRACT
The synthesis of a series of alpha-methylene-gamma-butyrolactones (compounds 4a, 4b, 6-12, 16, 17) and alpha,beta-unsaturated-delta-lactones (compounds 19-23, 25, 26) starting from 4,4-dimethyldihydrofuran-2,3-dione (1) has been described. Their chemical structures were assigned by spectroscopic evidence. These new compounds exhibited significantly different antiproliferative properties against cultured human tumor cell lines with their IC(50) values ranging from 0.88 to>20.00 microM.
Sigma-Aldrich
α-Methylene-γ-butyrolactone, 97%