Merck
  • Inhibitory effect of demoxepam on tryptophan binding to rat hepatic nuclei.
Inhibitory effect of demoxepam on tryptophan binding to rat hepatic nuclei.
Biochemical medicine and metabolic biology (1992-06-01)
H Sidransky, E Verney, J W Cosgrove, A M Schwartz
ABSTRACT
Since some patients with eosinophilia-myalgia syndrome ingested tryptophan along with benzodiazepines, we investigated whether demoxepam, the N-desalkylated compound of chlordiazepoxide, would influence the binding of tryptophan to hepatic nuclei. L-Tryptophan has been shown to bind (saturable, stereospecific, and of high affinity) to rat hepatic nuclei and nuclear envelopes. We report that demoxepam has an inhibitory effect on in vitro [3H]tryptophan binding to rat hepatic nuclei and has an apparent KD approximately 22 microM.
Product Description
Supelco
Demoxepam solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®