Merck
  • Oxidation process induced by 2-mercaptopyrimidine at a mercury electrode.
Oxidation process induced by 2-mercaptopyrimidine at a mercury electrode.
Journal of colloid and interface science (2005-06-15)
Josefa Angela García Calzón, Jesús Luis Muñiz Alvarez, Juan Miguel López Fonseca
ABSTRACT
The oxidation process induced by 2-mercaptopyrimidine (RSH) at a hanging mercury drop electrode was studied in aqueous media by normal and cyclic voltammetry. The results indicate the formation of a weakly adsorbed HgSR complex according to the reaction Hg+RSH-->HgSR+e(-)+H+. When the surface excess of HgSR is high, a two-dimensional condensation takes place if the potential is positive enough, as proved by phase-sensitive ac and cyclic voltammetry.
Sigma-Aldrich
2-Mercaptopyrimidine, 98%