Merck
  • Peripheral benzodiazepine receptor (PBR) ligand cytotoxicity unrelated to PBR expression.
Peripheral benzodiazepine receptor (PBR) ligand cytotoxicity unrelated to PBR expression.
Biochemical pharmacology (2005-02-16)
Gregory Hans, Sabine Wislet-Gendebien, François Lallemend, Pierre Robe, Bernard Rogister, Shibeshih Belachew, Laurent Nguyen, Brigitte Malgrange, Gustave Moonen, Jean-Michel Rigo
ABSTRACT
Some synthetic ligands of the peripheral-type benzodiazepine receptor (PBR), an 18 kDa protein of the outer mitochondrial membrane, are cytotoxic for several tumor cell lines and arise as promising chemotherapeutic candidates. However, conflicting results were reported regarding the actual effect of these drugs on cellular survival ranging from protection to toxicity. Moreover, the concentrations needed to observe such a toxicity were usually high, far above the affinity range for their receptor, hence questioning its specificity. In the present study, we have shown that micromolar concentrations of FGIN-1-27 and Ro 5-4864, two chemically unrelated PBR ligands are toxic for both PBR-expressing SK-N-BE neuroblastoma cells and PBR-deficient Jurkat lymphoma cells. We have thereby demonstrated that the cytotoxicity of these drugs is unrelated to their PBR-binding activity. Moreover, Ro 5-4864-induced cell death differed strikingly between both cell types, being apoptotic in Jurkat cells while necrotic in SK-N-BE cells. Again, this did not seem to be related to PBR expression since Ro 5-4864-induced death of PBR-transfected Jurkat cells remained apoptotic. Taken together, our results show that PBR is unlikely to mediate all the effects of these PBR ligands. They however confirm that some of these ligands are very effective cytotoxic drugs towards various cancer cells, even for reputed chemoresistant tumors such as neuroblastoma, and, surprisingly, also for PBR-lacking tumor cells.
Sigma-Aldrich
Anti-Mouse IgG (Fab specific)–Peroxidase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
N,N-Dihexyl-2-(4-fluorophenyl)indole-3-acetamide