Merck
  • Extramembrane central pore of multidrug exporter AcrB in Escherichia coli plays an important role in drug transport.
Extramembrane central pore of multidrug exporter AcrB in Escherichia coli plays an important role in drug transport.
The Journal of biological chemistry (2003-10-25)
Satoshi Murakami, Norihisa Tamura, Asami Saito, Takahiro Hirata, Akihito Yamaguchi
ABSTRACT
We previously reported the crystal structure of the major multidrug exporter AcrB in Escherichia coli (Murakami, S., Nakashima, R., Yamashita, E., and Yamaguchi, A. (2002) Nature 419, 587-593). The extramembrane headpiece of the AcrB trimer contains a central pore composed of three alpha-helices. Each pore helix belongs to a different monomer. In this study, we constructed cysteine-scanning mutants as to the residues comprising the pore helix. Of the 21 mutants (D99C to P119C), 5 (D101C, V105C, N109C, Q112C, and P116C) showed significantly reduced drug resistance and drug-exporting activity. These residues are localized on one side of the pore helix, i.e. on the wall of the pore. These observations strongly indicate the important role of this pore in the drug transport process. A N-ethylmaleimide binding experiment revealed that the pore is in the closed state, and the thickness of the permeability barrier in the middle of the pore corresponds to 2.5 alpha-helical turns. Two mutants (V105C and Q112C), which showed the greatest loss of activity of all of the pore mutants, were detected in the form of disulfide cross-linking dimers under normal conditions, suggesting that a conformational change of the pore is indispensable during the transport process.
Sigma-Aldrich
2-[4-(Dimethylamino)styryl]-1-ethylpyridinium iodide, ≥99% (HPLC), solid
Pricing and availability is not currently available.