Merck
  • Origin of Ca2+ necessary for carbachol-induced contraction in longitudinal muscle of the proximal colon of rats.
Origin of Ca2+ necessary for carbachol-induced contraction in longitudinal muscle of the proximal colon of rats.
Japanese journal of pharmacology (2002-02-07)
T Takeuchi, M Sumiyoshi, M Kitayama, N Hirayama, A Fujita, F Hata
ABSTRACT
The origin of Ca2+ necessary for carbachol (CCh)-induced contraction of longitudinal muscle of the proximal colon of rats was studied. CCh induced contraction of the muscle consisting of two phases, phasic and tonic phases, with a concomitant biphasic increase in [Ca2+]i. After removal of Ca2+ from the bathing solution of the colonic segments, CCh-induced contraction was rapidly inhibited; there was almost complete inhibition 1 min after the removal. Nicardipine, a blocker of voltage-dependent calcium channel, also significantly inhibited CCh-induced contraction. On the other hand, treatment of the colonic segments with thapsigargin, an inhibitor of sarcoplasmic reticulum (SR) Ca2+-ATPase, did not significantly affect the contraction except causing a slight decrease in the rate of contraction. These results suggest that Ca> entering through voltage-dependent calcium channels, but not released from SR, is essential for CCh-induced contraction of longitudinal muscle of the proximal colon of rats. This strict dependency of the CCh-induced contraction on extracellular Ca2+ was discussed in relation to the results obtained in the fundus of rats.
Sigma-Aldrich
ONO-RS-082, ≥97% (HPLC)