Merck
  • Prediabetic insulin resistance is not permissive to the development of cardiac resistance to insulin-like growth factor I in ventricular myocytes.
Prediabetic insulin resistance is not permissive to the development of cardiac resistance to insulin-like growth factor I in ventricular myocytes.
Diabetes research and clinical practice (2002-02-14)
Kadon K Hintz, Jun Ren
ABSTRACT
Resistance to insulin-like growth factor I (IGF-1)-induced cardiac contractile response has been reported in diabetes. To evaluate the role of prediabetic insulin resistance to cardiac IGF-1 resistance, whole body insulin resistance was generated with dietary sucrose and contractile function was evaluated in ventricular myocytes. Mechanical properties were evaluated using an IonOptix system and intracellular Ca(2+) transients were measured as changes in fura-2 fluorescence intensity (Delta FFI). After 8 weeks of feeding, sucrose rats displayed euglycemia, hepatomeglay and normal heart size, and glucose intolerance, confirming the presence of insulin resistance. Myocytes from sucrose-fed rats displayed decreased peak shortening (PS), reduced resting FFI, increased intracellular Ca(2+) clearing, associated with normal duration of shortening and relengthening compared to myocytes from starch-fed rats. IGF-1 (10(-10)-10(-6) M) caused a similar concentration-dependent decrease in PS in both groups. Only the highest concentration of IGF-1 elicited an inhibition on Delta FFI in sucrose myocytes. In addition, the IGF-1-induced response was abolished by the IGF-1 receptor antagonist H-1356 in both groups, and by the nitric oxide synthase inhibitor L-NAME in starch but not sucrose myocytes. These results indicated prediabetic insulin resistance alters cardiac contractile function at the myocytes level, but may not be permissive to cardiac contractile resistance to IGF-1.
Sigma-Aldrich
JB1 trifluoroacetate salt, ≥98% (HPLC)