Merck
  • Urocortin II enhances contractility in rabbit ventricular myocytes via CRF(2) receptor-mediated stimulation of protein kinase A.
Urocortin II enhances contractility in rabbit ventricular myocytes via CRF(2) receptor-mediated stimulation of protein kinase A.
Cardiovascular research (2006-01-03)
Li-Zhen Yang, Jens Kockskämper, Frank R Heinzel, Michael Hauber, Stefanie Walther, Joachim Spiess, Burkert Pieske
ABSTRACT
Urocortin II (UcnII), a peptide of the corticotropin-releasing factor (CRF) family, exerts profound actions on the cardiovascular system. Direct effects of UcnII on adult cardiomyocytes have not been evaluated before. Our aim was to characterize functional effects of UcnII on cardiomyocytes and to elucidate the underlying signaling pathway(s) and cellular mechanisms. Rabbit ventricular cardiomyocytes were stimulated at 0.5 Hz (22-25 degrees C). Unloaded cell shortening (FS, edge detection), [Ca(2+)](i) transients (Fluo-4), and L-type Ca(2+) currents (I(Ca), whole-cell patch clamping) were measured. Sarcoplasmic reticulum (SR) Ca(2+) load was assessed by rapid application of caffeine (20 mmol/L). UcnII increased cell shortening and accelerated relaxation in a time- and concentration-dependent manner (EC(50): 10.7 nmol/L). The inotropic effect of UcnII was maximal at 100 nmol/L (35%+/-11% increase in FS, n=8, P<0.05). The inotropic and lusitropic actions of UcnII were largely eliminated by inhibition of CRF(2) receptors (10 nmol/L antisauvagine-30, n=5) or protein kinase A (PKA, 500 nmol/L H-89, n=5). UcnII increased [Ca(2+)](i) transient amplitude (by 63%+/-35%, n=7, P<0.05) and decreased the time constant for decay (from 800+/-63 to 218+/-27 ms, n=7, P<0.001). UcnII also increased SR Ca(2+) load (by 19%+/-7%, n=7, P<0.05) and fractional Ca(2+) release (from 57%+/-7% to 98%+/-2%, n=7, P<0.01). I(Ca) was augmented by 32.7%+/-10.0% (n=9, P<0.05) and the I(Ca)-V relationship was shifted by -15 mV during UcnII treatment. UcnII exerts positive inotropic and lusitropic effects in cardiomyocytes via CRF(2) receptor-mediated stimulation of PKA which augments I(Ca) and SR Ca(2+) load to increase SR Ca(2+) release and [Ca(2+)](i) transients.
Sigma-Aldrich
Urocortin II mouse