- Supramolecular shuttle based on inclusion complex between cucurbit[6]uril and bispyridinium ethylene.
Supramolecular shuttle based on inclusion complex between cucurbit[6]uril and bispyridinium ethylene.
Organic letters (2011-11-10)
Viktor Kolman, Muhammad S A Khan, Martin Babinský, Radek Marek, Vladimir Sindelar
PMID22066799
ABSTRACT
Cucurbit[6]uril (CB6) and bispyridinium ethylene form a stable inclusion complex. A rotaxane derived from this complex was prepared in which a CB6 wheel shuttles along an axle in an NMR time-resolved regime.