Merck
  • Quantum chemical and vibrational investigation of sodium exchanged gamma-alumina surfaces.
Quantum chemical and vibrational investigation of sodium exchanged gamma-alumina surfaces.
Physical chemistry chemical physics : PCCP (2007-05-18)
Mathieu Digne, Pascal Raybaud, Philippe Sautet, Denis Guillaume, Hervé Toulhoat
ABSTRACT
The sodium cation is well known as an efficient poison of gamma-alumina surface acidity. This poisoning effect has been revealed both by characterization methods and catalytic tests. In this work, we propose an accurate model of sodium exchanged gamma-alumina surfaces. On realistic models of hydroxylated gamma-alumina surfaces, the location of sodium cation is determined by the use of density functional theory (DFT) methods. For the (100) and (110) surfaces of gamma-alumina, the sodium cation is found in a solvated state within an inner solvation sphere complex. Its coordination sphere is constituted by O-mu(2), O-mu(3) and HO-mu(1) surface groups. The stretching frequency of these HO-mu(1) groups is shifted, leading to the appearance of a new band predicted and observed at about 3754 cm(-1) on the IR spectrum.
Sigma-Aldrich
Sodium oxide, 80%