Merck
  • The gallium-deferoxamine complex: stability with different deferoxamine concentrations and incubation conditions.
The gallium-deferoxamine complex: stability with different deferoxamine concentrations and incubation conditions.
Applied radiation and isotopes : including data, instrumentation and methods for use in agriculture, industry and medicine (1998-09-24)
C Caraco, L Aloj, W C Eckelman
ABSTRACT
Previous studies report that deferoxamine (DFO) binds metallic ions such as Fe3+, In3+ and Ga3+ with very high affinity. This property of DFO has been utilized to label DFO-coupled compounds with radiometals such as 67Ga and 111In. We have studied the effect of low DFO concentrations and of different incubation conditions on the stability of the 67Ga-DFO complex. In our experience high (> 5 microM) DFO concentration appears to be critical in obtaining high radiochemical purity of such complexes.
Sigma-Aldrich
Gallium(III) chloride, anhydrous, beads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Gallium(III) chloride, anhydrous, beads, ≥99.999% trace metals basis