Merck
  • The effect of flavonoids on ofloxacin-induced mutagenicity in Euglena gracilis.
The effect of flavonoids on ofloxacin-induced mutagenicity in Euglena gracilis.
Mutation research (1998-09-03)
L Krizková, M Nagy, J Polónyi, L Ebringer
ABSTRACT
The antimutagenicity of 14 naturally occurring flavonoids (20 mumol/l) on ofloxacin (43 mumol/l and 86 mumol/l)-induced bleaching (mutagenicity) was studied in Euglena gracilis. The flavonoids chrysin, techtochrysin, chrysin-5-methylether galangin, galangin-5-methylether, pinocembrin and pinobanksin possess considerable antimutagenic properties against ofloxacin-induced bleaching of E. gracilis. Apigenin and isalpinin had only weak antimutagenic potency. Pinobanksin-5-methylether and pinobanksin-3-acetate showed very weak or no antimutagenic effect. However, kempferol, quercetin-3-methylether and quercetin-3,3'-dimethylether showed co-mutagenic or no antimutagenic effect depending on the concentration of ofloxacin. Two possible modes of action of the flavonoids on ofloxacin-induced bleaching of E. gracilis are discussed.
Sigma-Aldrich
Pinobanksin, ≥95% (HPLC)