Merck
  • Influence of a 1 h immobilization stress on sleep states and corticotropin-like intermediate lobe peptide (CLIP or ACTH18-39, Ph-ACTH18-39) brain contents in the rat.
Influence of a 1 h immobilization stress on sleep states and corticotropin-like intermediate lobe peptide (CLIP or ACTH18-39, Ph-ACTH18-39) brain contents in the rat.
Brain research (1997-03-14)
C Bonnet, L Léger, V Baubet, G Debilly, R Cespuglio
ABSTRACT
A 1 h immobilization stress (IS) was imposed to rats at the beginning of the dark period, i.e., when the animals start to be active. The IS was accompanied by an intense polygraphic waking and followed, over 12 h of the dark period, by a significant rebound of slow-wave sleep (SWS, +17%) and paradoxical sleep (PS, +57%). In order to estimate the IS-related changes in the endogenous concentrations of corticotropin-like intermediate lobe peptide (CLIP, ACTH18-39) and related compounds, a specific radioimmunoassay (RIA) was used. Assays performed in cerebral biopsies taken from arcuate (AN) and raphe dorsalis (nRD) nuclei led to the obtention of 2 main immunoreactive peaks, corresponding to CLIP and its phosphorylated form Ph-CLIP. Just after end of the IS and within the nRD. Ph-CLIP immunoreactivity increased by about 95%. Four hours later, i.e., when PS rebound was maximal, a 37% increase in Ph-CLIP immunoreactivity was measured in the AN. These observations have never been described before. In the blood, at the end of the restraint, CLIP/ACTH1-39 total immunoreactivity was increased by 330%. It returned to baseline level 4 h later. Blood concentration of corticosterone was also increased by 56% at the end of the IS and was close to baseline level 4 h later. Data reported here indicate that the IS first triggers an increase in Ph-CLIP within the nRD. Since the nRD contains sleep permissive components, this increase might be determinant for the SWS and PS rebound induction. The changes observed in the blood as regards CLIP/ACTH1-39 total immunoreactivity and corticosterone concentration testify to the efficacy of the IS and are part of the conventional picture accompanying such a situation. Finally, the increase in Ph-CLIP, occurring in the AN 4 h after the end of the restraint, might be part of the restorative processes necessary to compensate the stress overshoot.
Sigma-Aldrich
ProteoMass ACTH Fragment 18-39 MALDI-MS Standard, vial of 10 nmol, monoisotopic mol wt 2,464.1989 Da
Sigma-Aldrich
Adrenocorticotropic Hormone Fragment 18-39 human, ≥97% (HPLC)