Merck
  • Laccase-mediated formation of the phenoxazinone derivative, cinnabarinic acid.
Laccase-mediated formation of the phenoxazinone derivative, cinnabarinic acid.
FEBS letters (1995-12-04)
C Eggert, U Temp, J F Dean, K E Eriksson
ABSTRACT
The phenoxazinone chromophore occurs in a variety of biological systems, including numerous pigments and certain antibiotics. It also appears to form as part of a mechanism to protect mammalian tissue from oxidative damage. During cultivation of the basidiomycete, Pycnoporus cinnabarinus, a red pigment was observed to accumulate in the culture medium. It was identified as the phenoxazinone derivative, cinnabarinic acid (CA). Laccase was the predominant extracellular phenoloxidase activity in P. cinnabarinus cultures. In vitro studies showed that CA was formed after oxidation of the precursor, 3-hydroxyanthranilic acid (3-HAA), by laccases. Moreover, oxidative coupling of 3-HAA to form CA was also demonstrated for the mammalian counterpart of laccase, the blue copper oxidase, ceruloplasmin.
Sigma-Aldrich
Cinnabarinic Acid, ≥98% (HPLC)