Merck
  • Conformationally restricted congeners of dopamine derived from 2-aminoindan.
Conformationally restricted congeners of dopamine derived from 2-aminoindan.
Journal of medicinal chemistry (1982-12-01)
J G Cannon, J A Perez, R K Bhatnagar, J P Long, F M Sharabi
ABSTRACT
Two series of N-substituted 2-aminoindan systems have been prepared: 4,5-dihydroxy-2-aminoindan (1) has a hydroxylation pattern analogous to the alpha conformer of dopamine, and 5,6-dihydroxy-2-aminoindan (2) has a hydroxylation pattern of the beta conformer of dopamine. All members of both series demonstrated only extremely weak binding to calf caudate homogenate. Certain N-alkylated 4,5-dihydroxyindans were violent emetics in the dog and were potent in blockade of the effect of stimulation of the cardioaccelerator nerve of the cat. In contrast, the 5,6-dihydroxy series displayed low or no activity/potency in these assays. Conformational analysis of the 2-aminoindan system is described and discussed.
Sigma-Aldrich
2-Aminoindan hydrochloride, 98%