Merck
  • Studies on components of the chloroform-methanol extract from Madurella grisea.
Studies on components of the chloroform-methanol extract from Madurella grisea.
Journal of medical and veterinary mycology : bi-monthly publication of the International Society for Human and Animal Mycology (1986-04-01)
A E Cury, T Ioneda, P S Minami
ABSTRACT
Some of the lipids described in many fungi were found in Madurella grisea, in addition to other diethyl ether-soluble components with peculiar characteristics. One fraction structurally related to ethane diol (ethylene glycol) accounted for about 41% and a compound identified as methyl terephthalate represented about 38% of the extract. No steroids were detected. The absence of these lipids may perhaps explain the resistance of this organism to polyene antibiotics.
Sigma-Aldrich
mono-Methyl terephthalate, 97%