Merck
  • Liquid chromatographic methods for assay of carbamazepine, 10,11-dihydrocarbamazepine, and related compounds in carbamazepine drug substance and tablets.
Liquid chromatographic methods for assay of carbamazepine, 10,11-dihydrocarbamazepine, and related compounds in carbamazepine drug substance and tablets.
Journal - Association of Official Analytical Chemists (1987-09-01)
T D Cyr, F Matsui, R W Sears, N M Curran, E G Lovering
ABSTRACT
Liquid chromatographic (LC) methods have been developed for the determination of carbamazepine, the impurity 10,11-dihydrocarbamazepine, and related compounds in carbamazepine drug substance and tablets. The LC methods specify a 5 micron diol column and a mobile phase of acetonitrile-methanol-0.05% aqueous acetic acid (5 + 5 + 90). Iminodibenzyl and iminostilbene, starting materials for some routes of synthesis, elute late in the LC system; therefore, a thin-layer chromatographic method for their detection at the 0.05% level has been developed. Eight tablet and 13 raw material samples from several sources were examined. The impurities most frequently found were 10, 11-dihydrocarbamazepine and a compound identified as 10-bromocarbamazepine at levels up to 1.3 and 0.5%, respectively; minimum detectable amounts were about 0.01 and 0.03%, respectively.
Product Description
Carbamazepine impurity A, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
10,11-Dihydrocarbamazepine, 99%