Merck
  • Metabolic activation of phenacetin and phenetidine by several forms of cytochrome P-450 purified from liver microsomes of rats and hamsters.
Metabolic activation of phenacetin and phenetidine by several forms of cytochrome P-450 purified from liver microsomes of rats and hamsters.
Japanese journal of cancer research : Gann (1987-02-01)
T Nohmi, K Mizokami, S Kawano, M Fukuhara, M Ishidate
ABSTRACT
Metabolic activation of phenacetin by liver microsomes proceeds via both phenetidine and N-hydroxyphenacetin to direct-acting mutagens, i.e., N-hydroxyphenetidine and p-nitrosophenetole. Five different molecular species of cytochrome P-450 have been purified from liver microsomes of drug-pretreated Wistar rats or Syrian hamsters and their abilities to activate phenetidine and phenacetin were compared using reconstituted microsome systems. High-spin forms of cytochrome P-450 purified from 3-methylcholanthrene-pretreated rats (MC-P-448-H) or hamsters (P-488 ham-II) showed higher catalytic activity for N-hydroxylation of phenetidine than three other low-spin forms of cytochrome P-450 purified from the same animals or from phenobarbital-pretreated rats. MC-P-448-H and P-488 ham-II required the presence of cytochrome b5 for their maximum activities in the reconstituted system. The five forms of cytochrome P-450, however, exhibited no measurable activity for N-hydroxylation of phenacetin either with or without cytochrome b5. The mutagenicity of phenacetin and phenetidine toward Salmonella typhimurium TA100 was generated when the reconstituted microsomes containing MC-P-488-H or P-488 ham-II were used as activating enzymes. From these results, it was suggested that high-spin forms of cytochrome P-450 (MC-P-448-H and P-448 ham-II) played an important role in the metabolic activation of phenacetin to the direct-acting mutagens.
Sigma-Aldrich
4-Ethoxyaniline, 98%