Merck
  • Atomically dispersed Au-(OH)x species bound on titania catalyze the low-temperature water-gas shift reaction.
Atomically dispersed Au-(OH)x species bound on titania catalyze the low-temperature water-gas shift reaction.
Journal of the American Chemical Society (2013-02-27)
Ming Yang, Lawrence F Allard, Maria Flytzani-Stephanopoulos
ABSTRACT
We report a new method for stabilizing appreciable loadings (~1 wt %) of isolated gold atoms on titania and show that these catalyze the low-temperature water-gas shift reaction. The method combines a typical gold deposition/precipitation method with UV irradiation of the titania support suspended in ethanol. Dissociation of H2O on the thus-created Au-O-TiO(x) sites is facile. At higher gold loadings, nanoparticles are formed, but they were shown to add no further activity to the atomically bound gold on titania. Removal of this "excess" gold by sodium cyanide leaching leaves the activity intact and the atomically dispersed gold still bound on titania. The new materials may catalyze a number of other reactions that require oxidized active metal sites.
Sigma-Aldrich
Sodium cyanide, reagent grade, 97%
Sigma-Aldrich
Sodium cyanide, ACS reagent, ≥95.0%