- RS-WebPredictor: a server for predicting CYP-mediated sites of metabolism on drug-like molecules.
RS-WebPredictor: a server for predicting CYP-mediated sites of metabolism on drug-like molecules.
Bioinformatics (Oxford, England) (2012-12-18)
Jed Zaretzki, Charles Bergeron, Tao-wei Huang, Patrik Rydberg, S Joshua Swamidass, Curt M Breneman
PMID23242264
ABSTRACT
Regioselectivity-WebPredictor (RS-WebPredictor) is a server that predicts isozyme-specific cytochrome P450 (CYP)-mediated sites of metabolism (SOMs) on drug-like molecules. Predictions may be made for the promiscuous 2C9, 2D6 and 3A4 CYP isozymes, as well as CYPs 1A2, 2A6, 2B6, 2C8, 2C19 and 2E1. RS-WebPredictor is the first freely accessible server that predicts the regioselectivity of the last six isozymes. Server execution time is fast, taking on average 2s to encode a submitted molecule and 1s to apply a given model, allowing for high-throughput use in lead optimization projects. RS-WebPredictor is accessible for free use at http://reccr.chem.rpi.edu/Software/RS-WebPredictor/