Merck
  • Exposure of Carcinus maenas to waterborne fluoranthene: accumulation and multibiomarker responses.
Exposure of Carcinus maenas to waterborne fluoranthene: accumulation and multibiomarker responses.
The Science of the total environment (2012-12-12)
A P Rodrigues, K K Lehtonen, L Guilhermino, L Guimarães
ABSTRACT
Fluoranthene (FLU) is a priority polycyclic aromatic hydrocarbon (PAH) commonly detected in estuarine sediments, water and biota. Despite this, information on FLU detection, accumulation and effects on marine crustaceans is scarce. This work investigated the accumulation of FLU in Carcinus maenas and the responses of several early-warning biomarkers after a 7-day laboratory exposure to five FLU concentrations (2.56 to 100 μg L(-1)). After exposure to FLU, sub-samples of the crabs' digestive gland and muscle were collected for biomarker determinations. The remaining digestive gland and muscle, together with the rest of the whole-body soft tissues, were analysed for FLU residues by gas chromatography-mass spectrometry (GC-MS). The biomarkers assessed were: i) the quantification of FLU-type compounds by fixed wavelength fluorescence (FF); ii) the activities of glutathione S-transferases (GST) and glutathione reductase (GR), and the levels of total glutathione (GT) and lipid peroxidation (LPO) for oxidative stress; iii) the activity of acetylcholinesterase (AChE) for neurotoxicity; iv) the activities of isocitrate dehydrogenase (IDH) and lactate dehydrogenase (LDH) enzymes, and total protein, glycogen and lipids as indicators of changes in energy metabolism and storage; and v) the lysosomal membrane stability (LMS) as a measure of cell damage. The results showed strong (R(2)>0.95) concentration-dependent accumulation of FLU residues (as measured by GC-MS) in the remaining whole-body soft tissues and of FLU-type compounds (as measured by FF) in the digestive gland and muscle. A strong positive linear relationship (R(2)=0.91) between FLU residues and FLU-type compounds was also found. Comparing to controls, activities of GST and GR were significantly higher in crabs exposed to ≥16 and ≥40 μg L(-1) FLU, respectively. TG levels and IDH activity showed a significant trend to increase with FLU concentrations whereas AChE activity exhibited the opposite trend. FF measurements in the digestive gland and muscle proved to be an expeditious cost-effective method to assess the uptake and availability of FLU and its metabolites in C. maenas. The results suggest that under continuous environmental exposure, FLU may enhance detoxification and anti-oxidant defences, and cause alterations in the aerobic energy pathway, as well as neuromuscular toxic effects that may increase C. maenas risk of predation.
Sigma-Aldrich
Fluoranthene, 98%