Merck
  • Enhanced photovoltaic properties of Nb₂O₅-coated TiO₂ 3D ordered porous electrodes in dye-sensitized solar cells.
Enhanced photovoltaic properties of Nb₂O₅-coated TiO₂ 3D ordered porous electrodes in dye-sensitized solar cells.
ACS applied materials & interfaces (2012-11-17)
Hye-Na Kim, Jun Hyuk Moon
ABSTRACT
This paper describes the use of Nb₂O₅-coated TiO₂ 3D ordered porous electrodes in dye-sensitized solar cells. We employed bilayer inverse opal structures as a backbone of 3D porous structures, and the number of Nb₂O₅ coatings was controlled, determining the concentration of Nb₂O₅ coating. XPS measurements confirmed the formation of Nb₂O₅. The uniformity of the Nb₂O₅ coating was characterized by elemental mapping using SEM and TEM measurements. Photovoltaic measurement on dye-sensitized solar cells (DSSCs) that incorporated Nb₂O₅/TiO₂ inverse opal electrodes yielded a maximum efficiency of 7.23% for a 3.3 wt % Nb₂O₅ coating on a TiO₂ IO structure. The Nb₂O₅ significantly increased the short-circuit current density (J(SC)). Electrochemical impedance spectroscopy was used to measure the J(SC), revealing an enhanced electron injection upon deposition of the Nb₂O₅ coating.
Sigma-Aldrich
Niobium(V) oxide, −325 mesh, 99.9% trace metals basis
Sigma-Aldrich
Niobium(V) oxide, 99.99% trace metals basis