- Fluoridated HAp:Ln3+ (Ln = Eu or Tb) nanoparticles for cell-imaging.
Fluoridated HAp:Ln3+ (Ln = Eu or Tb) nanoparticles for cell-imaging.
Nanoscale (2012-10-19)
Junfeng Hui, Xiaoyong Zhang, Zhicheng Zhang, Shiqi Wang, Lei Tao, Yen Wei, Xun Wang
PMID23076865
ABSTRACT
Water-dispersible hydrophilic fluoridated HAp:Ln(3+) (Ln = Eu or Tb) nanoparticles were prepared via hydrophobic/hydrophilic transformation with surfactants (Pluronic F127). The HAp:Ln(3+) (Ln = Eu or Tb) nanoparticles with unique luminescent properties and excellent biocompatibility are promising for cell imaging applications.