Merck
  • Facile and durable antimicrobial finishing of cotton textiles using a silver salt and UV light.
Facile and durable antimicrobial finishing of cotton textiles using a silver salt and UV light.
Carbohydrate polymers (2012-10-10)
Marek Kozicki, Elżbieta Sąsiadek, Marek Kołodziejczyk, Justyna Komasa, Agnieszka Adamus, Waldemar Maniukiewicz, Aleksandra Pawlaczyk, Małgorzata Szynkowska, Jacek Rogowski, Edward Rybicki
ABSTRACT
In this study, we present facile antimicrobial finishing of cotton textiles. Screen printing was used for surface-finishing of cotton using a printing paste containing silver nitrate. UVC irradiation was applied to convert silver nitrate into a color product, thus also changing the color of the textiles. The color, its strength and stability of samples, depend on absorbed UVC energy and the formula of the printing paste. Scanning electron microscopy with the energy dispersive X-ray spectrometry revealed formation of silver particles on cotton threads; X-ray diffraction analysis and the time-of-flight secondary ion mass spectrometry did not provide clear information on these products. Microbiological studies revealed that the samples inhibited proliferation of Escherichia coli, Bacillus subtilis and Staphylococcus aureus. Washing fastness tests confirmed resistance of the samples to at least 50 washings. Additionally, the inhibition zones increased as the number of washing cycles increased, which is unique for such samples. This work also presents an approach to the design of antimicrobially finished workwear.
Sigma-Aldrich
Silver nitrate, ACS reagent, ≥99.0%
Sigma-Aldrich
Silver nitrate, puriss. p.a., ≥99.5% (AT)
Sigma-Aldrich
Silver nitrate, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8%
Sigma-Aldrich
Silver nitrate, BioXtra, >99% (titration)
Sigma-Aldrich
Silver nitrate, BioReagent, suitable for plant cell culture, >99% (titration)
Sigma-Aldrich
Silver nitrate, 99.9999% trace metals basis
Sigma-Aldrich
Silver nitrate, ReagentPlus®, ≥99.0% (titration)
Sigma-Aldrich
Silver nitrate, meets analytical specification of Ph. Eur., BP, USP, 99.8-100.5%
Sigma-Aldrich
Silver nitrate, tested according to Ph. Eur.
Sigma-Aldrich
Silver nitrate solution, 2.5 % (w/v) AgNO3 in H2O
Sigma-Aldrich
Silver nitrate on silica gel, extent of labeling: ~10 wt. % loading, +230 mesh