Merck
  • Long-term efficacy and safety comparison of liraglutide, glimepiride and placebo, all in combination with metformin in type 2 diabetes: 2-year results from the LEAD-2 study.
Long-term efficacy and safety comparison of liraglutide, glimepiride and placebo, all in combination with metformin in type 2 diabetes: 2-year results from the LEAD-2 study.
Diabetes, obesity & metabolism (2012-09-19)
M Nauck, A Frid, K Hermansen, A B Thomsen, M During, N Shah, T Tankova, I Mitha, D R Matthews
ABSTRACT
To investigate efficacy and safety of dual therapy with liraglutide and metformin in comparison to glimepiride and metformin, and metformin monotherapy over 2 years in patients with type 2 diabetes. In the 26-week the Liraglutide Effect and Action in Diabetes (LEAD)-2 core trial, patients (n = 1091) were randomized (2 : 2 : 2 : 1: 2) to liraglutide (0.6, 1.2 or 1.8 mg once-daily), placebo or glimepiride; all with metformin. Patients were enrolled if they were 18-80 years old with HbA1c 7.0-11.0% (previous monotherapy ≥3 months), or 7.0-10.0% (previous combination therapy ≥3 months), and body mass index ≤40 kg/m(2) . Patients completing the 26-week double-blinded phase could enter an 18-month open-label extension. HbA1c decreased significantly with liraglutide (0.4% with 0.6 mg, 0.6% with 1.2 and 1.8 mg) versus 0.3% increase with metformin monotherapy (p < 0.0001). HbA1c decrease with liraglutide was non-inferior versus 0.5% decrease with glimepiride. Liraglutide groups experienced significant weight loss (2.1, 3.0 and 2.9 kg with 0.6, 1.2 and 1.8 mg, respectively) compared to weight gain (0.7 kg) with glimepiride (p < 0.0001). Weight loss with liraglutide 1.2 and 1.8 mg was significantly greater than with metformin monotherapy (1.8 kg; p = 0.0185 and p = 0.0378 for 1.2 and 1.8 mg, respectively). The occurrence of minor hypoglycaemia was <5.0% in all liraglutide groups, significantly less than with glimepiride (24.0%; p < 0.0001). Liraglutide was well tolerated overall: gastrointestinal events were more common than with glimepiride or metformin monotherapy, but occurrence decreased with time. Liraglutide provided sustained glycaemic control over 2 years comparable to that provided by glimepiride. Liraglutide was well tolerated, and was associated with weight loss and a low rate of hypoglycaemia.
Sigma-Aldrich
Glimepiride, ≥98% (HPLC), solid
Glimepiride for system suitability, European Pharmacopoeia (EP) Reference Standard
USP
Glimepiride, United States Pharmacopeia (USP) Reference Standard
Glimepiride, European Pharmacopoeia (EP) Reference Standard